Learn More
Meso Scale Discovery R-PLEX Human IGFBP2 ASY
SDP
Supplier:  Meso Scale Discovery K151AAVR2
Encompass
The Meso Scale Discovery R-PLEX Human IGFBP-2 Assay is a multiplex immunoassay panel that measures Insulin-like Growth Factor Binding Protein 2 (IGFBP-2) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 MESO QuickPlex SQ 120 and V-PLEX Plus The R-PLEX Human IGFBP-2 Assay provides quantitative determination of IGFBP-2 an important regulator of Insulin-like Growth Factors
Catalog No. 50-000-79431