Learn More
Meso Scale Discovery R-PLEX Human Ikaros ASYQP
SDP
Supplier:  Meso Scale Discovery K1516UR22
Encompass
The R-PLEX Human Ikaros Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows for the simultaneous quantitative measurement of Ikaros protein in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance The assay has a typical lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL for Ikaros
Catalog No. 50-000-81714