Learn More
Meso Scale Discovery R-PLEX Human IL-17RB ASYQP
SDP
Supplier:  Meso Scale Discovery K151ACUR22
Encompass
The R-PLEX Human IL-17RB Assay is a multiplex immunoassay kit that enables simultaneous measurement of IL-17RB in human serum plasma and cell culture supernatants It utilizes Meso Scale Discovery electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It allows quantification of IL-17RB levels with accuracy and precision The kit has been validated for research use only and is not intended for diagnostic procedures
Catalog No. 50-000-79580