Learn More
Meso Scale Discovery R-PLEX Human IL-18BP ASYQP
SDP
Supplier:  Meso Scale Discovery K151J5R22
Encompass
The Meso Scale Discovery R-PLEX Human IL-18BP Assay is a multiplex immunoassay that quantitatively measures human IL-18BP in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the simultaneous measurement of IL-18BP with other biomarkers in human samples It can be used to monitor IL-18BP levels in inflammatory diseases or during immunotherapies that impact IL-18 signaling
Catalog No. 50-000-79545