Learn More
Meso Scale Discovery R-PLEX Human IL-36 ASYQP
SDP
Supplier:  Meso Scale Discovery K151S8R22
Encompass
The R-PLEX Human IL-36gamma Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures IL-36gamma in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78760