Learn More
Meso Scale Discovery R-PLEX Human Jagged-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AMCR22
Encompass
The R-Plex Human Jagged-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Jagged-1 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a sensitivity of 0 3 ng/mL and requires 25 L sample size The kit has been validated on the MESO QuickPlex SQ 120 instrument
Catalog No. 50-000-78737