Learn More
Meso Scale Discovery R-PLEX Human LBP ASY
SDP
Supplier:  Meso Scale Discovery K151K5R2
Encompass
The Meso Scale Discovery R-PLEX Human LBP Assay is a multiplex immunoassay panel that measures Lipopolysaccharide Binding Protein (LBP) in human serum plasma and cell culture supernatant This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and accurate way to quantify LBP levels with a detection range of 0 04-50 ng/mL The assay requires 50 L or less sample volume has high reproducibility and low non-specific binding The R-PLEX Human LBP Assay provides a simple and rapid way to measure this important biomarker
Catalog No. 50-000-82342