Learn More
Meso Scale Discovery R-PLEX Human LIF ASYQP
SDP
Supplier:  Meso Scale Discovery K151AJPR22
Encompass
The R-PLEX Human LIF Assay Sector 5-PL Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human leukemia inhibitory factor (LIF) in serum plasma and cell culture supernatants The kit contains a 96-well plate pre-coated with capture antibodies specific for human LIF along with detection antibodies and calibrators The electrochemiluminescent assay allows sensitive and accurate quantification of LIF levels with a detection range of 0 06-1500 pg/mL and requires only 25 L of sample This assay kit provides a high-throughput method for analyzing LIF levels in biological samples
Catalog No. 50-000-81627