Learn More
Meso Scale Discovery R-PLEX Human MAdCAM-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AAYR22
Encompass
The R-PLEX Human MAdCAM-1 Assay is an ELISA-based assay kit for quantifying MAdCAM-1 in human serum plasma and cell culture supernatants It contains a 96-well plate pre-coated with capture antibody detection antibody standards controls substrate and detailed instructions The assay has a detection range of 31 3-2000 pg/mL and takes 3 5 hours to complete It can be used to measure MAdCAM-1 levels for research purposes The kit includes sufficient reagents for one 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-79587