Learn More
Meso Scale Discovery R-PLEX Human Mesothelin ASYQP
SDP
Supplier:  Meso Scale Discovery K1510HR22
Encompass
The R-PLEX Human Mesothelin Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures levels of mesothelin in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to run the assay Results are obtained in as little as 3 5 hours This assay can be used for cancer biomarker research and monitoring therapeutic response
Catalog No. 50-000-80552