Learn More
Meso Scale Discovery R-PLEX Human MMP-13 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AJSR22
Encompass
The R-Plex Human MMP-13 Assay from Meso Scale Diagnostics quantifies human MMP-13 in various samples using electrochemiluminescence (ECL) technology It offers high sensitivity broad range low sample use and quick results The kit includes a 96-well plate with capture and ECL-labeled detection antibodies calibrators controls diluent buffers seals and instructions It tests up to 80 samples in duplicates in 3 5 hours with a detection limit of 19 pg/mL ensuring sensitive and accurate MMP-13 quantification
Catalog No. 50-000-81665