Learn More
Meso Scale Discovery R-PLEX Human MMP-3 ASYQP
SDP
Supplier:  Meso Scale Discovery K151ZFR22
Encompass
The R-PLEX Human MMP-3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures levels of active and pro-MMP-3 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and sample diluent The assay has a typical lower limit of detection of 0 03 ng/mL and an upper limit of 10 000 ng/mL
Catalog No. 50-000-80111