Learn More
Meso Scale Discovery R-PLEX Human Nesfatin-1 ASY
SDP
Supplier:  Meso Scale Discovery K151ABVR2
Encompass
The Meso Scale Discovery R-PLEX Human Nesfatin-1 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Nesfatin-1 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of Nesfatin-1 levels This assay has high sensitivity wide dynamic range and requires low sample volume The R-PLEX assay format allows simultaneous measurement of Nesfatin-1 and other biomarkers in the same sample This assay is for research use only and not for use in diagnostic procedures
Catalog No. 50-000-80324