Learn More
Meso Scale Discovery R-PLEX Human Neuropilin-1 ASY
SDP
Supplier:  Meso Scale Discovery K151ABWR2
Encompass
The R-PLEX Human Neuropilin-1 Assay is a multiplex immunoassay kit for measuring Neuropilin-1 and up to 45 other biomarkers It uses ECL detection on the SECTOR S 600 instrument requiring only 25 L of sample and a 2 5-hour run time The kit includes a plate with capture antibodies detection antibodies calibrators controls wash buffers signal reagents and consumables for 96 tests It has a sensitivity of 0 3 ng/mL and uses serum plasma (EDTA) or cell culture supernatants enabling high-throughput sensitive and reproducible quantification of Neuropilin-1
Catalog No. 50-000-78939