Learn More
Meso Scale Discovery R-PLEX Human NPTX-1 ASY
SDP
Supplier:  Meso Scale Discovery K151T8R2
Encompass
The Meso Scale Discovery R-PLEX Hu Neuronal Pentraxin 1 Assay is a multiplex immunoassay kit for measuring human Neuronal Pentraxin 1 (NP1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L or less of sample per well and takes about 3 5 hours The kit includes a 96-well plate detection antibodies calibrators and controls measuring NP1 levels from 0 05 - 1000 ng/mL aiding in neurodegenerative disease research
Catalog No. 50-000-79166