Learn More
Meso Scale Discovery R-PLEX Human Osteonectin ASYQP
SDP
Supplier:  Meso Scale Discovery K151ZJR22
Encompass
The R-PLEX Human Osteonectin Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of osteonectin in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting osteonectin with a broad dynamic range This multiplex assay allows researchers to efficiently analyze osteonectin levels alongside other biomarkers relevant to bone metabolism and cancer progression With high sensitivity broad dynamic range and minimal sample volume requirements this assay serves as a useful tool for studying osteonectin in the laboratory
Catalog No. 50-000-80108