Learn More
Meso Scale Discovery R-PLEX Human PC9/PCSK9 ASYQP
SDP
Supplier:  Meso Scale Discovery K151ABAR22
Encompass
The R-PLEX Human PC9/PCSK9 Assay Kit from Meso Scale Diagnostics measures PCSK9 and PC9 in human serum plasma and cell culture supernatants using an ECL-based assay It employs MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation from a small sample Sensitivity is 0 3 ng/mL for PCSK9 and 15 pg/mL for PC9 The kit includes a 96-well plate antibodies calibrators controls buffers and seals enabling high-throughput analysis of cholesterol-related biomarkers
Catalog No. 50-000-79430