Learn More
Meso Scale Discovery R-PLEX Human PSGL-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151YHR22
Encompass
The R-PLEX Human PSGL-1 Assay Kit from Meso Scale Discovery measures Prostate Specific Membrane Antigen (PSMA) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with a PSMA capture antibody a SULFO-TAG labeled detection antibody diluents calibrators and controls It detects PSMA concentrations from 0 3 - 2500 pg/mL with high specificity and minimal cross-reactivity offering a robust tool for studying PSMA biology and therapies
Catalog No. 50-000-79822