Learn More
Meso Scale Discovery R-PLEX Human PSP-D ASYQP
SDP
Supplier:  Meso Scale Discovery K1519XR22
Encompass
The R-PLEX Human PSP-D Assay from Meso Scale Diagnostics is a multiplex immunoassay panel measuring Prostate Specific Protein (PSP) and three prostate cancer biomarkers (Free PSA Intact PSA Human Kallikrein-2) in serum and plasma Using MSD s MULTI-ARRAY technology it provides sensitive accurate results from a small sample The 4-plex panel aids in prostate cancer screening and monitoring The kit includes a 96-well plate with capture antibodies detection antibodies and MSD GOLD SULFO-TAG labels for high sensitivity
Catalog No. 50-000-78948