Learn More
Meso Scale Discovery R-PLEX Human Reg-3a ASYQP
SDP
Supplier:  Meso Scale Discovery K151U8R22
Encompass
The R-PLEX Human REG3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring REG3 in human serum plasma and cell culture supernatants This ECL assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation of REG3 with a detection range of 0 04 ng/mL to 10 000 pg/mL The kit includes a 96-well plate with capture and detection antibodies calibrators controls buffers and seals for one plate It enables high-throughput screening of REG3 levels with a 3 5-hour protocol
Catalog No. 50-000-78525