Learn More
Meso Scale Discovery R-PLEX Human S100A8/MRP8 ASYQP
SDP
Supplier:  Meso Scale Discovery K1510PR22
Encompass
The R-PLEX Human S100A8/MRP8 Assay Kit from Meso Scale Diagnostics measures S100A8/MRP8 protein levels in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents S100A8/MRP8 is a calcium-binding protein involved in inflammation useful in research and drug development
Catalog No. 50-000-80196