Learn More
Meso Scale Discovery R-PLEX Human SCF ASY
SDP
Supplier:  Meso Scale Discovery K1510QR2
Encompass
The R-Plex Human SCF Assay Kit from Meso Scale Discovery measures soluble stem cell factor (SCF) in human serum plasma cell culture supernatants and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of SCF The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and controls It has a sensitivity of 1 5 pg/mL and an upper limit of 1500 pg/mL enabling high-throughput SCF quantification for research
Catalog No. 50-000-78383