Learn More
Meso Scale Discovery R-PLEX Human Syndecan-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151V4R22
Encompass
The Meso Scale Discovery R-PLEX Human Syndecan-1 Assay is a multiplex immunoassay panel that measures Syndecan-1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of Syndecan-1 which plays a role in cell adhesion migration proliferation angiogenesis and tumorigenesis It requires 50 L of sample and can perform up to 80 tests across two plates The kit includes all necessary reagents and plates for sample testing
Catalog No. 50-000-79123