Learn More
Meso Scale Discovery R-PLEX Human TGM-2 ASY
SDP
Supplier:  Meso Scale Discovery K151AKCR2
Encompass
The R-PLEX Human TGM-2 Assay Kit from Meso Scale Discovery measures tissue transglutaminase (TGM-2) in human serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a dynamic range of up to 4 logs enabling high-throughput and accurate TGM-2 quantification
Catalog No. 50-000-78868