Learn More
Meso Scale Discovery R-PLEX Human TNFAIP6/TSG-6 ASY
SDP
Supplier:  Meso Scale Discovery K1510UR2
Encompass
The R-PLEX Human TNF IP6/TSG-6 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures TNF -induced protein 6 (TSG-6) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and Streptavidin-Gold The assay can quantify TSG-6 between 0 04 - 10 000 pg/mL with an incubation time of 2 hours
Catalog No. 50-000-82417