Learn More
Meso Scale Discovery R-PLEX Human tPA ASY
SDP
Supplier:  Meso Scale Discovery K151ACZR2
Encompass
The R-PLEX Human TPA Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures tissue plasminogen activator (TPA) in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for quantifying TPA levels The assay has a detection range of 3 05-10 000 pg/mL and exhibits minimal cross-reactivity with related molecules Each kit provides reagents for one 96-well plate which can process 40 samples in duplicate This assay enables high-throughput quantitative analysis of TPA which plays a key role in fibrinolysis and thrombosis
Catalog No. 50-000-80363