Learn More
Meso Scale Discovery R-PLEX Human TSP-1 ASY
SDP
Supplier:  Meso Scale Discovery K1519FR2
Encompass
The R-PLEX Human TSP-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the concentration of Thrombospondin-1 (TSP-1) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Sample Diluent and Calibrator The assay can detect TSP-1 in the range of 0 05-500 ng/mL and requires only 25 L of sample
Catalog No. 50-000-79446