Learn More
Meso Scale Discovery R-PLEX Human VE-Cadherin ASYQP
SDP
Supplier:  Meso Scale Discovery K151AJVR22
Encompass
The R-Plex Human VE-Cadherin Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human Vascular Endothelial (VE)-cadherin in cell and tissue lysates It uses the Meso Scale Discovery electrochemiluminescence (ECL) technology and provides a rapid ultra-sensitive and high-throughput method for measuring VE-cadherin levels The assay requires only 25 L of sample per well and can detect VE-cadherin at sub-picogram levels The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators and assay diluent It allows high-throughput profiling of VE-cadherin across multiple samples
Catalog No. 50-000-81621