Learn More
Meso Scale Discovery R-PLEX Human Visfatin ASYQP
SDP
Supplier:  Meso Scale Discovery K151Q5R22
Encompass
The Meso Scale Discovery R-PLEX Human Visfatin Assay is a multiplex immunoassay panel that measures the adipokine visfatin in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The visfatin assay is available as part of a custom multiplex panel or as an individual plex The assay requires 50 L of sample and can be read on MSD instrument platforms including SECTOR Imagers and QuickPlex
Catalog No. 50-000-81667