Learn More
Meso Scale Discovery R-PLEX Human VTCN1/B7-H4 ASY
SDP
Supplier:  Meso Scale Discovery K151ALMR2
Encompass
The R-PLEX Human VTCN1 (B7-H4) Assay is a multiplex immunoassay panel that measures the concentration of VTCN1 (B7-H4) in human serum plasma (EDTA heparin) and cell culture supernatant samples This assay uses electrochemiluminescence (ECL) detection technology on the Meso Scale Discovery (MSD) platform The assay has a detection range of 0 61 - 2500 pg/mL and includes calibrators and controls to ensure accurate quantitation This assay allows high-throughput sensitive and reproducible quantification of VTCN1 (B7-H4) levels from small sample volumes It is intended for research use only to study VTCN1 (B7-H4) biology and its role in diseases like cancer
Catalog No. 50-000-79657