Learn More
Meso Scale Discovery R-PLEX Mouse IL-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K1521YR22
Encompass
The R-PLEX Mouse IL-1alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that allows for the simultaneous quantitative measurement of mouse interleukin 1 alpha (IL-1 ) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The assay can detect mouse IL-1 with a lower limit of detection of 0 05 pg/mL and an upper limit of 10 000 pg/mL The total assay time is 3 5 hours Each kit provides sufficient reagents to perform 1 x 96-well plate when used as directed
Catalog No. 50-000-82147