Learn More
Meso Scale Discovery R-PLEX Mouse MIG ASY
SDP
Supplier:  Meso Scale Discovery K1520IR2
Encompass
The R-PLEX Mouse MIG Assay from Meso Scale Discovery is a multiplex immunoassay measuring mouse IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 and IL-17 in serum plasma and cell culture supernatants This 10-plex assay uses MSD s electrochemiluminescent detection technology offering high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies read buffer calibrators and controls Results are available in 3 5 hours using MSD s SECTOR Imager
Catalog No. 50-000-80509