Learn More
Meso Scale Discovery R-PLEX Mouse TIMP-1 ASY
SDP
Supplier:  Meso Scale Discovery K152YOR2
Encompass
The R-PLEX Mouse TIMP-1 Assay is a multiplex immunoassay panel that measures TIMP-1 in mouse samples such as serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The panel allows simultaneous measurement of TIMP-1 along with other biomarkers This assay requires the SECTOR Imager 6000 and is available as a kit with reagents and consumables for 96 tests
Catalog No. 50-000-79681