Learn More
Meso Scale Discovery R-PLEX Mouse TIMP-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K152YOR22
Encompass
The Meso Scale Discovery R-PLEX Mouse TIMP-1 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of mouse TIMP-1 in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology on patterned arrays for high sensitivity and a wide dynamic range The assay requires 50 L or less of sample and can be read on the MESO QuickPlex SQ 120 instrument The assay is available as a singleplex or as part of a multiplex panel This assay allows sensitive reproducible quantitation of mouse TIMP-1 for life science research applications
Catalog No. 50-000-82263