Learn More
Meso Scale Discovery R-PLEX Mouse VCAM-1 ASY
SDP
Supplier:  Meso Scale Discovery K1527PR2
Encompass
The R-PLEX Mouse VCAM-1 Assay Kit from Meso Scale Discovery measures VCAM-1 in mouse samples like serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate antibodies buffers blocker solutions calibrator and controls It detects VCAM-1 from 1 95-1000 pg/mL with minimal dilution and takes 3 5 hours This aids research on VCAM-1 in inflammation cardiovascular disease cancer and more
Catalog No. 50-000-82204