Learn More
Meso Scale Discovery R-PLEX Rat Adiponectin ASYQP
SDP
Supplier:  Meso Scale Discovery K152YQR22
Encompass
The Meso Scale Discovery R-PLEX Rat Adiponectin Assay Kit is designed for the quantitative measurement of adiponectin in rat serum plasma and tissue culture supernatants using an electrochemiluminescent detection technology The assay has a detection range of 78-50 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The kit includes reagents for one 96-well plate and can perform up to 40 tests in duplicate The assay requires a Meso Scale Discovery SECTOR Imager for plate reading
Catalog No. 50-000-79648