Learn More
Meso Scale Discovery R-PLEX Rat EPO ASYQP
SDP
Supplier:  Meso Scale Discovery K1531GR22
Encompass
The R-PLEX Rat EPO Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying erythropoietin (EPO) in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive rapid EPO detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 0 3 pg/mL with an upper limit of 1500 pg/mL Results are available in 3 5 hours offering a fast sensitive method for EPO quantification
Catalog No. 50-000-81877