Learn More
Meso Scale Discovery R-PLEX Rat IL-10 ASYQP
SDP
Supplier:  Meso Scale Discovery K1531VR22
Encompass
The R-PLEX Rat IL-10 Assay from Meso Scale Discovery measures rat interleukin 10 (IL-10) in various biological samples using MSD s electrochemiluminescent detection technology It offers high sensitivity a wide dynamic range and requires low sample volume The assay quantifies IL-10 concentrations from 0 05-1000 pg/mL with high specificity and minimal cross-reactivity The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents for rapid and accurate IL-10 quantification in research
Catalog No. 50-000-82390