Learn More
Meso Scale Discovery R-PLEX Rat IL-13 ASYQP
SDP
Supplier:  Meso Scale Discovery K1531WR22
Encompass
The R-PLEX Rat IL-13 Assay from Meso Scale Discovery is a multiplex immunoassay that measures rat interleukin 13 (IL-13) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less of sample per well and can be read on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 MESO QuickPlex SQ 120 and V-PLEX Plus The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for rat IL-13
Catalog No. 50-000-82482