Learn More
Meso Scale Discovery R-PLEX Rat IL-1B ASYQP
SDP
Supplier:  Meso Scale Discovery K1531ZR22
Encompass
The R-PLEX Rat IL-1beta Assay from Meso Scale Discovery is a multiplex immunoassay for measuring rat interleukin-1 beta (IL-1beta) in various biological samples It uses MSD s electrochemiluminescence detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay quantifies IL-1beta concentrations from 0 05-1000 pg/mL with a 2-hour incubation It includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation enabling rapid accurate IL-1beta measurement for research
Catalog No. 50-000-78679