Learn More
Meso Scale Discovery R-PLEX Rat MCP-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K1531DR22
Encompass
The R-PLEX Rat MCP-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures rat monocyte chemoattractant protein 1 (MCP-1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82238