Learn More
Meso Scale Discovery R-PLEX SARS-CoV-2 Spike ASY
SDP
Supplier:  Meso Scale Discovery K150ADBR2
Encompass
The R-PLEX SARS-CoV-2 Spike Assay is a multiplex immunoassay panel that detects antibodies to the SARS-CoV-2 spike protein receptor binding domain (RBD) and nucleocapsid protein (N) It uses electrochemiluminescence (ECL) on the Meso Scale Discovery (MSD) platform for high sensitivity and broad dynamic range This assay can help determine recent or prior infection with SARS-CoV-2 It includes calibrators and controls and has a testing time of approximately 2 5 hours for 96 wells The R-PLEX SARS-CoV-2 Spike Assay kit provides reagents for one 96-well plate
Catalog No. 50-000-82283