Learn More
Medchemexpress LLC (R)-praziquantel-d11 | 1399880-38-6 | MFCD00005008 | 99.6% | 323.47 | C19H13D11N2O2 | 1MG
SDP
Supplier:  Medchemexpress LLC HY126057S1MG
Encompass_Preferred
(R)-Praziquantel-d11 is the deuterium-labeled R-enantiomer of praziquantel supplied as a research-grade analytical standard. It is intended for use as an internal standard in mass spectrometry and for laboratory studies of the active enantiomer's pharmacology; it is supplied in milligram quantities and is not for human or clinical use.
  • Deuterium-labeled R-enantiomer (d11) for analytical use.
  • Suitable as an internal standard for mass spectrometry.
  • High chemical purity (99.6%) and defined molecular weight (323.47).
  • Molecular formula C19H13D11N2O2.
  • Supplied in small-mass formats for research use only.
Catalog No. 50-002-36227