Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R-praziquantel-d11 | 1399880-38-6 | MFCD19705058 | 99.6% | 323.47 g·mol⁻¹ | C19H13D11N2O2 | 5MG
SDP

Supplier:  Medchemexpress LLC HY126057S5MG

Encompass_Preferred

(R)-Praziquantel-d11 is the deuterium-labeled R-enantiomer of praziquantel supplied as a high-purity analytical standard for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS workflows. The material is provided as a solid with recommended storage conditions to preserve isotopic and chemical integrity.

  • Deuterium-labeled R-enantiomer of praziquantel.
  • High chemical purity (≈99.6%).
  • Formula: C19H13D11N2O2; molecular weight ≈323.5 g·mol⁻¹.
  • Supplied as a solid suitable for preparation of analytical standards.
  • Intended use as an internal standard or tracer for NMR, GC-MS, and LC-MS quantitation.
  • Recommended storage: powder at -20°C for long term; solutions at -80°C for extended storage.

Catalog No. 50-002-36481


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.