Learn More
Medchemexpress LLC (R)-Propranolol (hydrochloride) | 13071-11-9 | 99.3% | C16H22ClNO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0295100MG
Encompass_Preferred
(R)-Propranolol hydrochloride is a less active enantiomer of the nonselective β-adrenergic receptor (βAR) antagonist propranolol. It exhibits high affinity for β1AR and β2AR, and is intended for research use only.
  • Less active enantiomer of propranolol
  • Nonselective β-adrenergic receptor (βAR) antagonist
  • Exhibits high affinity for β1AR and β2AR
  • Can prevent adrenaline-induced cardiac arrhythmias
  • Capable of reversing ventricular tachycardia caused by ouabain
Catalog No. 50-004-32229