Learn More
Medchemexpress LLC (R)-pyrrolidine-2-carboxylic acid | 344-25-2 | ≥98.0% | C5H9NO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY750875G
Encompass_Preferred
(R)-pyrrolidine-2-carboxylic acid is a proline isomer that demonstrates renal and hepatotoxicity in rats. It is utilized in studies of amino acid metabolism and toxicity mechanisms, presenting as a white to off-white solid.
  • Supports research into amino acid metabolism
  • Aids in the study of toxicity mechanisms
  • Characterized as an endogenous metabolite
  • Exhibits solubility in water and DMSO
Catalog No. 50-004-34806