Learn More
Medchemexpress LLC (R)-Pyrrolidine-3-carboxylic acid | 72580-54-2 | 97.0% | C5H9NO2 | 25 G

Supplier: Medchemexpress LLC 25GHY60256
(R)-Pyrrolidine-3-carboxylic acid is a proline derivative. Amino acids and their derivatives are used as ergogenic supplements that influence anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and help prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Proline derivative
- Influences anabolic hormone secretion
- Influences fuel supply during exercise
- Influences mental performance under stress
- Helps prevent exercise-induced muscle damage
- Beneficial ergogenic dietary substance
- Solid, white to light yellow appearance
- Purity of 97.0%
- Soluble in H2O: 20 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.