Learn More
Medchemexpress LLC (R,R)-GSK321 | 1816272-19-1 | 99.4% | 501.55 g·mol⁻¹ | C28H28FN5O3 | 5 MG

Supplier: Medchemexpress LLC HY1288885MG
(R,R)-GSK321 is the (R,R) enantiomer of GSK321, a research-grade inhibitor of isocitrate dehydrogenase 1 (IDH1) with reported activity against both wild-type and R132H mutant forms. Supplied as a high-purity solid, it is intended for biochemical and cellular research into IDH1-related pathways and cancer models.
- Target IDH1 inhibitor with reported cellular activity.
- Molecular formula C28H28FN5O3; molecular weight 501.55 g·mol⁻¹.
- Purity 99.4%.
- Physical form solid, white to off-white powder; suitable for storage at low temperature.
- High solubility in DMSO; prepare stock solutions as recommended.
- Provided for research use only in biochemical and cellular assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.