Learn More
Medchemexpress LLC (R,R)-(+)-Hydrobenzoin | 52340-78-0 | 99.8% | C14H14O2 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY5912510MM/1ML
Encompass_Preferred
(R,R)-(+)-Hydrobenzoin is a chiral secondary alcohol that serves as a key substrate or a chiral auxiliary reagent in various catalytic reactions and synthetic experiments. It is intended for research use only.
  • Can be used as a chiral template in oxoselenation reactions for synthesizing chiral cyclopentanol and aminocyclopentanol.
  • Can act as a substrate in deoxygenation and dehydration reactions catalyzed by a molybdenum (Mo) base, yielding trans-stilbene and benzaldehyde.
Catalog No. 50-004-33806