Learn More
Medchemexpress LLC (R,R)-(+)-Hydrobenzoin | 52340-78-0 | 99.8% | C14H14O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY591255G
Encompass_Preferred
(R,R)-(+)-Hydrobenzoin is a chiral secondary alcohol that can be used as a key substrate or a chiral auxiliary reagent in various catalytic reactions and synthetic experiments. It can serve as a chiral template in an oxoselenation reaction to asymmetrically functionalize cyclopentene, thereby synthesizing chiral cyclopentanol and aminocyclopentanol.
  • Acts as a substrate in deoxygenation and dehydration reactions.
  • Generates trans-stilbene and benzaldehyde when catalyzed by a molybdenum (Mo) base catalyst.
Catalog No. 50-004-34455